missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cPs

Catalog No. AA3671822
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3671822 100 g
AA3671836 500 g
AA36718A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3671822 Supplier Thermo Scientific Chemicals Supplier No. 03671822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Methyl cellulose is used as a thickener and an emulsifier in various food and cosmetic products; as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. In paper and textile industries, it is used as a sizing, thereby protecting the fibers from water or oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Quantity 100 g
Merck Index 14,6040
Solubility Information Soluble in cold water.
Physical Form Powder
Chemical Name or Material Methyl cellulose, Viscosity: 4000cPs
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only