missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 15 cP (2% solution in water)

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Supplier:  Thermo Scientific Chemicals 182310010

 View more versions of this product

Catalog No. AC182310010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray-White
Viscosity (2% Solution in Water) 12-18 CP
9004-67-5
MFCD00081763
1 kg
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
454.513
Authentic
Plastic bottle
Methyl cellulose
Powder
C20H38O11
15, 6111
YLGXILFCIXHCMC-JHGZEJCSSA-N
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
51063134
5% max.
12 to 18cP (2% solution in water,20°C)

Chemical Identifiers

9004-67-5
454.513
YLGXILFCIXHCMC-JHGZEJCSSA-N
(5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
C20H38O11
MFCD00081763
51063134
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : FJ5959000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only