missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 15 cP (2% solution in water)

Catalog No. p-161201
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182311000 100 g Plastic bottle
AC182312500 250 g Plastic bottle
AC182310010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC182311000 Supplier Thermo Scientific Chemicals Supplier No. 182311000
Only null left

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color Gray-White
Physical Form Powder
Merck Index 15, 6111
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic bottle
Viscosity 12 to 18cP (2% solution in water,20°C)
Chemical Name or Material Methyl cellulose, Viscosity (2% Solution in Water) 12-18 CP
RTECSNumber FJ5959000
TSCA TSCA

RUO – Research Use Only