missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Cellulose (20-30mPa.s, 2% in Water at 20°C), TCI America™
$56.95 - $159.49
Chemical Identifiers
| CAS | 9004-67-5 |
|---|---|
| Molecular Formula | C20H38O11 |
| Molecular Weight (g/mol) | 454.513 |
| MDL Number | MFCD00081763 |
| InChI Key | YLGXILFCIXHCMC-JHGZEJCSSA-N |
| PubChem CID | 51063134 |
| IUPAC Name | (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| SMILES | COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Chemical Identifiers
| 9004-67-5 | |
| 454.513 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| C20H38O11 | |
| MFCD00081763 | |
| 51063134 | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Specifications
| 9004-67-5 | |
| C20H38O11 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane | |
| 51063134 | |
| Methyl Cellulose (20-30mPa.s, 2% in Water at 20°C) |
| White | |
| MFCD00081763 | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC | |
| 454.513 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (8)-0186
RTECSNumber : FJ5959000
TSCA : Yes