Learn More
Methyl caffeate
Inhibitor of protein tyrosine kinase | CAS: 3843-74-1 | C10H10O4 | 194.186 g/mol
Supplier: Thermo Scientific Chemicals J6378603
Description
Inhibitor of protein tyrosine kinase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl caffeate | |
| 158°C to 161°C | |
| C10H10O4 | |
| 1 g | |
| methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 | |
| COC(=O)C=CC1=CC(=C(C=C1)O)O | |
| 194.186 | |
| CHEBI:6856 | |
| Powder |
| 3843-74-1 | |
| Yellow | |
| MFCD00210468 | |
| 14,1635 | |
| OCNYGKNIVPVPPX-HWKANZROSA-N | |
| methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate | |
| 689075 | |
| 194.19 |
Chemical Identifiers
| 3843-74-1 | |
| 194.186 | |
| OCNYGKNIVPVPPX-HWKANZROSA-N | |
| 689075 | |
| methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
| C10H10O4 | |
| MFCD00210468 | |
| methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 | |
| CHEBI:6856 | |
| COC(=O)C=CC1=CC(=C(C=C1)O)O |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only