missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Caffeate 98.0+%, TCI America™
$229.10 - $693.07
Chemical Identifiers
| CAS | 3843-74-1 |
|---|---|
| Molecular Formula | C10H10O4 |
| Molecular Weight (g/mol) | 194.186 |
| MDL Number | MFCD00210468 |
| InChI Key | OCNYGKNIVPVPPX-HWKANZROSA-N |
| Synonym | methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 |
| PubChem CID | 689075 |
| ChEBI | CHEBI:6856 |
| IUPAC Name | methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
| SMILES | COC(=O)C=CC1=CC(=C(C=C1)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2519200MG
|
TCI America
M2519200MG |
200 mg |
Each for $229.10
|
|
|||||
|
M25191G
|
TCI America
M25191G |
1 g |
Each for $693.07
|
|
|||||
Chemical Identifiers
| 3843-74-1 | |
| 194.186 | |
| OCNYGKNIVPVPPX-HWKANZROSA-N | |
| 689075 | |
| methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
| C10H10O4 | |
| MFCD00210468 | |
| methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 | |
| CHEBI:6856 | |
| COC(=O)C=CC1=CC(=C(C=C1)O)O |
Specifications
| 3843-74-1 | |
| C10H10O4 | |
| 200 mg | |
| OCNYGKNIVPVPPX-HWKANZROSA-N | |
| methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate | |
| 689075 | |
| 194.19 | |
| Crystalline Powder |
| 160°C | |
| MFCD00210468 | |
| methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001 | |
| COC(=O)C=CC1=CC(=C(C=C1)O)O | |
| 194.186 | |
| CHEBI:6856 | |
| ≥98.0% (GC,T) | |
| Methyl Caffeate |
Safety and Handling
TSCA : No