missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl behenate, 99%, analytical standard for GC
CAS: 929-77-1 | C23H46O2 | 354.62 g/mol
$112.34
Chemical Identifiers
| CAS | 929-77-1 |
|---|---|
| Molecular Formula | C23H46O2 |
| Molecular Weight (g/mol) | 354.62 |
| MDL Number | MFCD00009347 |
| InChI Key | QSQLTHHMFHEFIY-UHFFFAOYSA-N |
| Synonym | methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf |
| PubChem CID | 13584 |
| IUPAC Name | methyl docosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC226810010
|
N/A | 1 g |
Each for $112.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C23H46O2 | |
| MFCD00009347 | |
| methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf | |
| methyl docosanoate |
Specifications
| 929-77-1 | |
| Authentic | |
| Glass Bottle | |
| CH3(CH2)20CO2CH3 | |
| 15,1022 | |
| (5% in Hexane) Clear colorless solution | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)OC | |
| 354.62 | |
| 354.61 | |
| Methyl behenate |
| 54.0°C to 56.0°C | |
| 98.5% min. (GC) | |
| C23H46O2 | |
| MFCD00009347 | |
| methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf | |
| QSQLTHHMFHEFIY-UHFFFAOYSA-N | |
| methyl docosanoate | |
| 13584 | |
| 99% |
Safety and Handling
EINECSNumber : 213-207-8
RUO – Research Use Only