missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Arachidate 98.0+%, TCI America™
$151.89 - $457.57
Chemical Identifiers
| CAS | 1120-28-1 |
|---|---|
| Molecular Formula | C21H42O2 |
| Molecular Weight (g/mol) | 326.57 |
| MDL Number | MFCD00009014 |
| InChI Key | QGBRLVONZXHAKJ-UHFFFAOYSA-N |
| Synonym | methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate |
| PubChem CID | 14259 |
| IUPAC Name | methyl icosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCC(=O)OC |
Chemical Identifiers
| C21H42O2 | |
| MFCD00009014 | |
| methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate | |
| methyl icosanoate |
Specifications
| 1120-28-1 | |
| White | |
| C21H42O2 | |
| methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate | |
| CCCCCCCCCCCCCCCCCCCC(=O)OC | |
| 326.57 | |
| 326.57 | |
| Crystalline Powder |
| 48°C | |
| 369°C | |
| MFCD00009014 | |
| QGBRLVONZXHAKJ-UHFFFAOYSA-N | |
| methyl icosanoate | |
| 14259 | |
| ≥98.0% (GC) | |
| Methyl Arachidate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes