missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Arachidate 98.0+%, TCI America™
Supplier: TCI America A090025G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C21H42O2 | |
| MFCD00009014 | |
| methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate | |
| methyl icosanoate |
Specifications
| Methyl Arachidate | |
| 48°C | |
| 369°C | |
| 25 g | |
| methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate | |
| CCCCCCCCCCCCCCCCCCCC(=O)OC | |
| 326.57 | |
| 326.57 | |
| Crystalline Powder |
| 1120-28-1 | |
| White | |
| C21H42O2 | |
| MFCD00009014 | |
| QGBRLVONZXHAKJ-UHFFFAOYSA-N | |
| methyl icosanoate | |
| 14259 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes