missing translation for 'onlineSavingsMsg'
Learn More

Methyl Arachidate 98.0+%, TCI America™

Supplier:  TCI America A09005G

 View more versions of this product

Catalog No. A09005G


Only null left

Description

Description

Specifications

Specifications

Methyl Arachidate
48°C
369°C
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
CCCCCCCCCCCCCCCCCCCC(=O)OC
326.57
326.57
Crystalline Powder
1120-28-1
White
C21H42O2
5 g
QGBRLVONZXHAKJ-UHFFFAOYSA-N
methyl icosanoate
14259
≥98.0% (GC)

Chemical Identifiers

1120-28-1
326.57
QGBRLVONZXHAKJ-UHFFFAOYSA-N
14259
CCCCCCCCCCCCCCCCCCCC(=O)OC
C21H42O2
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
methyl icosanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions