missing translation for 'onlineSavingsMsg'
Learn More

Methyl adipoyl chloride, 96%

CAS: 35444-44-1 | C7H11ClO3 | 178.61 g/mol

Supplier:  Thermo Scientific Chemicals L0437709

 View more versions of this product

Catalog No. AAL0437709


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl adipoyl chloride
1.05
C7H11ClO3
CH3O2C(CH2)4COCl
10 g
1766187
hexanedioic acid monochloride monomethyl ester, hexanoic acid, 6-chloro-6-oxo-, methyl ester, 6-methoxy-6-oxohexanoyl chloride, methyl 5-chloroformyl pentanoate, 5-chlorocarbonyl-pentanoic acid methyl ester, mono-methyl adipoyl chloride, 5-carbomethoxyvaleryl chloride, methyl 6-chloro-6-oxocaproate, methyl adipoyl chloride
COC(=O)CCCCC(Cl)=O
178.61
178.62
35444-44-1
76°C (0.8 mmHg)
1.447
MFCD00013661
UN3265
Moisture sensitive
HDLGIEZOMYJKAK-UHFFFAOYSA-N
methyl 6-chloro-6-oxohexanoate
520737
96%

Chemical Identifiers

35444-44-1
178.61
HDLGIEZOMYJKAK-UHFFFAOYSA-N
520737
COC(=O)CCCCC(Cl)=O
C7H11ClO3
MFCD00013661
hexanedioic acid monochloride monomethyl ester, hexanoic acid, 6-chloro-6-oxo-, methyl ester, 6-methoxy-6-oxohexanoyl chloride, methyl 5-chloroformyl pentanoate, 5-chlorocarbonyl-pentanoic acid methyl ester, mono-methyl adipoyl chloride, 5-carbomethoxyvaleryl chloride, methyl 6-chloro-6-oxocaproate, methyl adipoyl chloride
methyl 6-chloro-6-oxohexanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 252-569-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only