missing translation for 'onlineSavingsMsg'
Learn More

Methyl adipoyl chloride, 96%

Catalog No. p-7044072
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0437704 2 g
AAL0437709 10 g
AAL0437718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0437704 Supplier Thermo Scientific Chemicals Supplier No. L0437704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35444-44-1 | C7H11ClO3 | 178.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35444-44-1
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.61
MDL Number MFCD00013661
InChI Key HDLGIEZOMYJKAK-UHFFFAOYSA-N
Synonym hexanedioic acid monochloride monomethyl ester, hexanoic acid, 6-chloro-6-oxo-, methyl ester, 6-methoxy-6-oxohexanoyl chloride, methyl 5-chloroformyl pentanoate, 5-chlorocarbonyl-pentanoic acid methyl ester, mono-methyl adipoyl chloride, 5-carbomethoxyvaleryl chloride, methyl 6-chloro-6-oxocaproate, methyl adipoyl chloride
PubChem CID 520737
IUPAC Name methyl 6-chloro-6-oxohexanoate
SMILES COC(=O)CCCCC(Cl)=O

Specifications

Density 1.05
Boiling Point 76°C (0.8 mmHg)
Refractive Index 1.447
Linear Formula CH3O2C(CH2)4COCl
Quantity 2 g
UN Number UN3265
Beilstein 1766187
Sensitivity Moisture sensitive
Formula Weight 178.62
Percent Purity 96%
Chemical Name or Material Methyl adipoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-569-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only