missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetoacetate 99.0+%, TCI America™

Catalog No. A065025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A065025G 25 g
A0650500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A065025G Supplier TCI America Supplier No. A065025G

Chemical Identifiers

CAS 105-45-3
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008784
InChI Key WRQNANDWMGAFTP-UHFFFAOYSA-N
Synonym methyl acetoacetate, acetoacetic acid methyl ester, methylacetoacetate, methyl 3-oxobutyrate, butanoic acid, 3-oxo-, methyl ester, methyl acetylacetate, methyl acetylacetonate, 3-oxobutanoic acid methyl ester, acetoacetic methyl ester, acetoacetic acid, methyl ester
PubChem CID 7757
IUPAC Name methyl 3-oxobutanoate
SMILES COC(=O)CC(C)=O

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 170°C
Quantity 25 g
Formula Weight 116.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Acetoacetate
EINECSNumber (2)-1474&(2)-1475&(2)-1505
RTECSNumber AK5775000
TSCA Yes