missing translation for 'onlineSavingsMsg'
Learn More

Methyl acetoacetate, 99%

Catalog No. AAA136960B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA136960B 1000 g
AAA1369630 250 g
AAA136960I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA136960B Supplier Thermo Scientific Chemicals Supplier No. A136960B
Only null left

CAS: 105-45-3 | C5H8O3 | 116.12 g/mol

Methyl acetoacetate is used as a chemical reagent used in the synthesis of pharmaceuticals. It participates in the Biginelli reaction, forming molecules including dihydropyrimidinones.Methyl acetoacetate (MAA) is used for the synthesis of alpha-substituted aceto- acetic esters and cyclic compounds, e.g. pyrazole, pyrimidine and coumarin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl acetoacetate is used as a chemical reagent used in the synthesis of pharmaceuticals. It participates in the Biginelli reaction, forming molecules including dihydropyrimidinones.Methyl acetoacetate (MAA) is used for the synthesis of alpha-substituted aceto- acetic esters and cyclic compounds, e.g. pyrazole, pyrimidine and coumarin derivatives.

Solubility
Soluble in water

Notes
Store at 2-8°C. Incompatible with oxidizing agents and bases. Protect from light.

Chemical Identifiers

CAS 105-45-3
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008784
InChI Key WRQNANDWMGAFTP-UHFFFAOYSA-N
Synonym methyl acetoacetate, acetoacetic acid methyl ester, methylacetoacetate, methyl 3-oxobutyrate, butanoic acid, 3-oxo-, methyl ester, methyl acetylacetate, methyl acetylacetonate, 3-oxobutanoic acid methyl ester, acetoacetic methyl ester, acetoacetic acid, methyl ester
PubChem CID 7757
IUPAC Name methyl 3-oxobutanoate
SMILES COC(=O)CC(C)=O

Specifications

Melting Point -80°C
Density 1.076
Boiling Point 170°C
Flash Point 70°C (158°F)
Odor Pleasant
Refractive Index 1.4185
Linear Formula CH3COCH2CO2CH3
Quantity 1000 g
Beilstein 506727
Merck Index 14,6009
Solubility Information Soluble in water
Formula Weight 116.12
Percent Purity 99%
Chemical Name or Material Methyl acetoacetate
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 203-299-8
RTECSNumber AK5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only