missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 7-bromoheptanoate, 98%
CAS: 54049-24-0 | C8H15BrO2 | 223.11 g/mol
$163.48 - $477.03
Chemical Identifiers
| CAS | 54049-24-0 |
|---|---|
| Molecular Formula | C8H15BrO2 |
| Molecular Weight (g/mol) | 223.11 |
| MDL Number | MFCD02258672 |
| InChI Key | BXRLUWIDTDLHQE-UHFFFAOYSA-N |
| Synonym | methyl-7-bromoheptanoate, 7-bromoheptanoic acid methyl ester, heptanoic acid, 7-bromo-, methyl ester, acmc-1aya7, 13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one |
| PubChem CID | 554086 |
| IUPAC Name | methyl 7-bromoheptanoate |
| SMILES | COC(=O)CCCCCCBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2723406
|
Thermo Scientific Chemicals
H2723406 |
5 g |
Each for $163.48
|
|
|||||
|
AAH2723414
|
Thermo Scientific Chemicals
H2723414 |
25 g |
Each for $477.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54049-24-0 | |
| 223.11 | |
| BXRLUWIDTDLHQE-UHFFFAOYSA-N | |
| 554086 | |
| COC(=O)CCCCCCBr |
| C8H15BrO2 | |
| MFCD02258672 | |
| methyl-7-bromoheptanoate, 7-bromoheptanoic acid methyl ester, heptanoic acid, 7-bromo-, methyl ester, acmc-1aya7, 13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one | |
| methyl 7-bromoheptanoate |
Specifications
| 54049-24-0 | |
| C8H15BrO2 | |
| 5 g | |
| BXRLUWIDTDLHQE-UHFFFAOYSA-N | |
| methyl 7-bromoheptanoate | |
| 554086 | |
| 98% |
| 1.257 | |
| MFCD02258672 | |
| methyl-7-bromoheptanoate, 7-bromoheptanoic acid methyl ester, heptanoic acid, 7-bromo-, methyl ester, acmc-1aya7, 13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one | |
| COC(=O)CCCCCCBr | |
| 223.11 | |
| 223.11 | |
| Methyl 7-bromoheptanoate |
Safety and Handling
TSCA : No
RUO – Research Use Only