CAS RN 54049-24-0
IUPAC Name:
methyl 7-bromoheptanoate
Synonyms:
methyl 7-bromoheptanoate
Molecular Weight (g/mol):
223.11
Molecular Formula:
C8H15BrO2
InChi Key:
BXRLUWIDTDLHQE-UHFFFAOYSA-N
SMILES:
COC(=O)CCCCCCBr
1
–
1
of
1
results
1
Promotions Available
Methyl 7-bromoheptanoate, 98%
Methyl 7-bromoheptanoate, 98%
CAS: 54049-24-0 Molecular Formula: C8H15BrO2 Molecular Weight (g/mol): 223.11 MDL Number: MFCD02258672 InChI Key: BXRLUWIDTDLHQE-UHFFFAOYSA-N Synonym: methyl-7-bromoheptanoate,7-bromoheptanoic acid methyl ester,heptanoic acid, 7-bromo-, methyl ester,acmc-1aya7,13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one PubChem CID: 554086 IUPAC Name: methyl 7-bromoheptanoate SMILES: COC(=O)CCCCCCBr