missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-bromoheptanoate, 98%

Catalog No. AAH2723414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2723414 25 g
AAH2723406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2723414 Supplier Thermo Scientific Chemicals Supplier No. H2723414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54049-24-0 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54049-24-0
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD02258672
InChI Key BXRLUWIDTDLHQE-UHFFFAOYSA-N
Synonym methyl-7-bromoheptanoate, 7-bromoheptanoic acid methyl ester, heptanoic acid, 7-bromo-, methyl ester, acmc-1aya7, 13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one
PubChem CID 554086
IUPAC Name methyl 7-bromoheptanoate
SMILES COC(=O)CCCCCCBr

Specifications

Density 1.257
Quantity 25 g
Formula Weight 223.11
Percent Purity 98%
Chemical Name or Material Methyl 7-bromoheptanoate

RUO – Research Use Only