Learn More
Methyl 6-chloropyrazine-2-carboxylate, 95%
CAS: 23611-75-8 | C6H5ClN2O2 | 172.568 g/mol
Supplier: Thermo Scientific Chemicals H6668203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl 6-chloropyrazine-2-carboxylate | |
| C6H5ClN2O2 | |
| 1 g | |
| methyl 6-chloro-2-pyrazinecarboxylate, 2-chloro-6-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid, 6-chloro-, methyl ester, 2-chloro-6-co2me-pyrazine, 6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 6-chloro-, methyl ester, pubchem16685, 6-chloro-pyrazine-2-carboxylicacidmethylester, ksc494k2p, methyl-6-chloropyrazine-2-carboxylate | |
| COC(=O)C1=CN=CC(=N1)Cl | |
| 172.568 | |
| 172.57 | |
| Solid |
| 23611-75-8 | |
| MFCD10686598 | |
| 880043 | |
| MVVYUJFEXRODQA-UHFFFAOYSA-N | |
| methyl 6-chloropyrazine-2-carboxylate | |
| 11084353 | |
| 95% |
Chemical Identifiers
| 23611-75-8 | |
| 172.568 | |
| MVVYUJFEXRODQA-UHFFFAOYSA-N | |
| 11084353 | |
| COC(=O)C1=CN=CC(=N1)Cl |
| C6H5ClN2O2 | |
| MFCD10686598 | |
| methyl 6-chloro-2-pyrazinecarboxylate, 2-chloro-6-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid, 6-chloro-, methyl ester, 2-chloro-6-co2me-pyrazine, 6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 6-chloro-, methyl ester, pubchem16685, 6-chloro-pyrazine-2-carboxylicacidmethylester, ksc494k2p, methyl-6-chloropyrazine-2-carboxylate | |
| methyl 6-chloropyrazine-2-carboxylate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only