missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-chloropyrazine-2-carboxylate, 95%

Catalog No. p-7057229
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66682MD 250 mg
AAH6668203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66682MD Supplier Thermo Scientific Chemicals Supplier No. H66682MD
Only null left

CAS: 23611-75-8 | C6H5ClN2O2 | 172.568 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23611-75-8
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD10686598
InChI Key MVVYUJFEXRODQA-UHFFFAOYSA-N
Synonym methyl 6-chloro-2-pyrazinecarboxylate, 2-chloro-6-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid, 6-chloro-, methyl ester, 2-chloro-6-co2me-pyrazine, 6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 6-chloro-, methyl ester, pubchem16685, 6-chloro-pyrazine-2-carboxylicacidmethylester, ksc494k2p, methyl-6-chloropyrazine-2-carboxylate
PubChem CID 11084353
IUPAC Name methyl 6-chloropyrazine-2-carboxylate
SMILES COC(=O)C1=CN=CC(=N1)Cl

Specifications

Quantity 250 mg
Beilstein 880043
Formula Weight 172.57
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 6-chloropyrazine-2-carboxylate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only