missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Phenylvalerate 98.0+%, TCI America™

Catalog No. P13245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13245G 5 g
P132425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13245G Supplier TCI America Supplier No. P13245G
Only null left

Chemical Identifiers

CAS 20620-59-1
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00025896
InChI Key BNNWLDUOVGYRLY-UHFFFAOYSA-N
Synonym 5-Phenylvaleric Acid Methyl Ester
PubChem CID 262942
IUPAC Name methyl 5-phenylpentanoate
SMILES COC(=O)CCCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 173°C
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Phenylvalerate