accessibility menu, dialog, popup

UserName

CAS RN 20620-59-1

H3COO

CAS RN 20620-59-1

IUPAC Name: methyl 5-phenylpentanoate
Synonyms: methyl 5-phenylpentanoate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: BNNWLDUOVGYRLY-UHFFFAOYSA-N
SMILES: COC(=O)CCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results