missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-hexynoate, 95%

Catalog No. p-3586451
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC380880050 5 g Glass bottle
AC380880250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC380880050 Supplier Thermo Scientific Chemicals Supplier No. 380880050
Only null left

CAS: 77758-51-1 | C7H10O2 | 126.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77758-51-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
MDL Number MFCD00671366
InChI Key LZULAZTXJLWELL-UHFFFAOYSA-N
Synonym methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester
PubChem CID 534077
IUPAC Name methyl hex-5-ynoate
SMILES COC(=O)CCCC#C

Specifications

Color Colorless to Yellow
Boiling Point 60°C to 62°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.435
Linear Formula CHC(CH2)3COOCH3
Quantity 5 g
Formula Weight 126.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 5-hexynoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only