missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Formylsalicylate 98.0+%, TCI America™
Supplier: TCI America M208225G
Specifications
| Methyl 5-Formylsalicylate | |
| 81°C | |
| C9H8O4 | |
| 25 g | |
| XJKWVPNWHOCFBR-UHFFFAOYSA-N | |
| methyl 5-formyl-2-hydroxybenzoate | |
| 14363598 | |
| ≥98.0% (GC,T) |
| 41489-76-3 | |
| Yellow | |
| MFCD03840784 | |
| 5-Formylsalicylic Acid Methyl Ester, Methyl 5-Formyl-2-hydroxybenzoate, 5-Formyl-2-hydroxybenzoic Acid Methyl Ester | |
| COC(=O)C1=C(O)C=CC(C=O)=C1 | |
| 180.16 | |
| 180.16 | |
| Crystalline Powder |
Chemical Identifiers
| 41489-76-3 | |
| 180.16 | |
| XJKWVPNWHOCFBR-UHFFFAOYSA-N | |
| 14363598 | |
| COC(=O)C1=C(O)C=CC(C=O)=C1 |
| C9H8O4 | |
| MFCD03840784 | |
| 5-Formylsalicylic Acid Methyl Ester, Methyl 5-Formyl-2-hydroxybenzoate, 5-Formyl-2-hydroxybenzoic Acid Methyl Ester | |
| methyl 5-formyl-2-hydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-4640
TSCA : No