CAS RN 41489-76-3
IUPAC Name:
methyl 5-formyl-2-hydroxybenzoate
Synonyms:
methyl 5-formyl-2-hydroxybenzoate
Molecular Weight (g/mol):
180.16
Molecular Formula:
C9H8O4
InChi Key:
XJKWVPNWHOCFBR-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(O)C=CC(C=O)=C1
1
–
1
of
1
results
1
Methyl 5-Formylsalicylate 98.0+%, TCI America™
CAS: 41489-76-3 Molecular Formula: C9H8O4 Molecular Weight (g/mol): 180.16 MDL Number: MFCD03840784 InChI Key: XJKWVPNWHOCFBR-UHFFFAOYSA-N Synonym: 5-Formylsalicylic Acid Methyl Ester, Methyl 5-Formyl-2-hydroxybenzoate, 5-Formyl-2-hydroxybenzoic Acid Methyl Ester PubChem CID: 14363598 IUPAC Name: methyl 5-formyl-2-hydroxybenzoate SMILES: COC(=O)C1=C(O)C=CC(C=O)=C1