missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Formylsalicylate 98.0+%, TCI America™

Catalog No. M20825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20825G 5 g
M208225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20825G Supplier TCI America Supplier No. M20825G

Chemical Identifiers

CAS 41489-76-3
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD03840784
InChI Key XJKWVPNWHOCFBR-UHFFFAOYSA-N
Synonym 5-Formylsalicylic Acid Methyl Ester, Methyl 5-Formyl-2-hydroxybenzoate, 5-Formyl-2-hydroxybenzoic Acid Methyl Ester
PubChem CID 14363598
IUPAC Name methyl 5-formyl-2-hydroxybenzoate
SMILES COC(=O)C1=C(O)C=CC(C=O)=C1

Specifications

Melting Point 81°C
Color Yellow
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Formylsalicylate
EINECSNumber (3)-4640
TSCA No