missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-chloro-5-oxovalerate, 97%

Catalog No. AAB2514109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2514109 10 g
AAB2514118 50 g
AAB2514130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2514109 Supplier Thermo Scientific Chemicals Supplier No. B2514109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1501-26-4 | C6H9ClO3 | 164.59 g/mol

Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases and moisture.

Chemical Identifiers

CAS 1501-26-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000756
InChI Key JCAZSWWHFJVFPP-UHFFFAOYSA-N
Synonym methyl 4-chloroformyl butyrate, methyl 5-chloro-5-oxovalerate, methyl 4-chloroformylbutyrate, methyl5-chloro-5-oxopentanoate, 5-chloro-5-oxopentanoic acid methyl ester, methyl glutaryl chloride, pentanoic acid, 5-chloro-5-oxo-, methyl ester, 4-chlorocarbonyl-butyric acid methyl ester, zlchem 559, pubchem14033
PubChem CID 73916
IUPAC Name methyl 5-chloro-5-oxopentanoate
SMILES COC(=O)CCCC(Cl)=O

Specifications

Boiling Point 108°C to 110°C (15 mmHg)
Refractive Index 1.447
Linear Formula ClCO(CH2)3CO2CH3
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material Methyl 5-chloro-5-oxovalerate
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-115-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only