accessibility menu, dialog, popup

UserName

H3COOClO

CAS RN 1501-26-4

IUPAC Name: methyl 5-chloro-5-oxopentanoate
Synonyms: methyl 5-chloro-5-oxopentanoate
Molecular Weight (g/mol): 164.59
Molecular Formula: C6H9ClO3
InChi Key: JCAZSWWHFJVFPP-UHFFFAOYSA-N
SMILES: COC(=O)CCCC(Cl)=O
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)