missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Chloro-2-nitrobenzoate 98.0+%, TCI America™
$29.83 - $48.46
Chemical Identifiers
| CAS | 51282-49-6 |
|---|---|
| Molecular Formula | C8H6ClNO4 |
| Molecular Weight (g/mol) | 215.589 |
| MDL Number | MFCD00272139 |
| InChI Key | JGBJHRKCUKTQOE-UHFFFAOYSA-N |
| Synonym | 5-chloro-2-nitrobenzoic acid methyl ester, benzoic acid, 5-chloro-2-nitro-, methyl ester, unii-1t5d4k1v51, methyl 5-chloro-2-nitro-benzoate, pubchem3712, acmc-209ksr, dsstox_cid_24948, dsstox_rid_80608, dsstox_gsid_44948, ksc495i7r |
| PubChem CID | 171018 |
| IUPAC Name | methyl 5-chloro-2-nitrobenzoate |
| SMILES | COC(=O)C1=C(C=CC(=C1)Cl)[N+](=O)[O-] |
Chemical Identifiers
| 51282-49-6 | |
| 215.589 | |
| JGBJHRKCUKTQOE-UHFFFAOYSA-N | |
| 171018 | |
| COC(=O)C1=C(C=CC(=C1)Cl)[N+](=O)[O-] |
| C8H6ClNO4 | |
| MFCD00272139 | |
| 5-chloro-2-nitrobenzoic acid methyl ester, benzoic acid, 5-chloro-2-nitro-, methyl ester, unii-1t5d4k1v51, methyl 5-chloro-2-nitro-benzoate, pubchem3712, acmc-209ksr, dsstox_cid_24948, dsstox_rid_80608, dsstox_gsid_44948, ksc495i7r | |
| methyl 5-chloro-2-nitrobenzoate |
Specifications
| 51282-49-6 | |
| C8H6ClNO4 | |
| 5 g | |
| JGBJHRKCUKTQOE-UHFFFAOYSA-N | |
| methyl 5-chloro-2-nitrobenzoate | |
| 171018 | |
| ≥98.0% (GC) | |
| Methyl 5-Chloro-2-nitrobenzoate |
| 50°C | |
| MFCD00272139 | |
| 5-chloro-2-nitrobenzoic acid methyl ester, benzoic acid, 5-chloro-2-nitro-, methyl ester, unii-1t5d4k1v51, methyl 5-chloro-2-nitro-benzoate, pubchem3712, acmc-209ksr, dsstox_cid_24948, dsstox_rid_80608, dsstox_gsid_44948, ksc495i7r | |
| COC(=O)C1=C(C=CC(=C1)Cl)[N+](=O)[O-] | |
| 215.589 | |
| 215.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes