CAS RN 51282-49-6
IUPAC Name:
methyl 5-chloro-2-nitrobenzoate
Synonyms:
methyl 5-chloro-2-nitrobenzoate
Molecular Weight (g/mol):
215.59
Molecular Formula:
C8H6ClNO4
InChi Key:
JGBJHRKCUKTQOE-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Cl)=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
Methyl 5-Chloro-2-nitrobenzoate 98.0+%, TCI America™
CAS: 51282-49-6 Molecular Formula: C8H6ClNO4 Molecular Weight (g/mol): 215.589 MDL Number: MFCD00272139 InChI Key: JGBJHRKCUKTQOE-UHFFFAOYSA-N Synonym: 5-chloro-2-nitrobenzoic acid methyl ester,benzoic acid, 5-chloro-2-nitro-, methyl ester,unii-1t5d4k1v51,methyl 5-chloro-2-nitro-benzoate,pubchem3712,acmc-209ksr,dsstox_cid_24948,dsstox_rid_80608,dsstox_gsid_44948,ksc495i7r PubChem CID: 171018 IUPAC Name: methyl 5-chloro-2-nitrobenzoate SMILES: COC(=O)C1=C(C=CC(=C1)Cl)[N+](=O)[O-]