missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Chloro-2-nitrobenzoate 98.0+%, TCI America™

Catalog No. M19965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19965G 5 g
M199625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19965G Supplier TCI America Supplier No. M19965G
Only null left

Chemical Identifiers

CAS 51282-49-6
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00272139
InChI Key JGBJHRKCUKTQOE-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrobenzoic acid methyl ester, benzoic acid, 5-chloro-2-nitro-, methyl ester, unii-1t5d4k1v51, methyl 5-chloro-2-nitro-benzoate, pubchem3712, acmc-209ksr, dsstox_cid_24948, dsstox_rid_80608, dsstox_gsid_44948, ksc495i7r
PubChem CID 171018
IUPAC Name methyl 5-chloro-2-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 50°C
Quantity 5 g
Formula Weight 215.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Chloro-2-nitrobenzoate