missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Bromovalerate 97.0+%, TCI America™

Catalog No. M22655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22655G 5 g
M226525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22655G Supplier TCI America Supplier No. M22655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5454-83-1
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000265
InChI Key RAVVJKCSZXAIQP-UHFFFAOYSA-N
PubChem CID 79557
IUPAC Name methyl 5-bromopentanoate
SMILES COC(=O)CCCCBr

Specifications

Color Yellow
Boiling Point 89°C
Quantity 5 g
Synonym methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate
Formula Weight 195.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Bromovalerate