missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromovalerate, 97%

Catalog No. p-161121
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126460100 10 g Glass bottle
AC126460250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC126460100 Supplier Thermo Scientific Chemicals Supplier No. 126460100
Only null left

CAS: 5454-83-1 | C6H11BrO2 | 195.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5454-83-1
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000265
InChI Key RAVVJKCSZXAIQP-UHFFFAOYSA-N
PubChem CID 79557
IUPAC Name methyl 5-bromopentanoate
SMILES COC(=O)CCCCBr

Specifications

Color Brown to Colorless
Density 1.3600g/mL
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4620 to 1.464
Linear Formula Br(CH2)4CO2CH3
Quantity 10 g
Specific Gravity 1.36
Synonym methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate
Formula Weight 195.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 5-bromovalerate, 97%
Show More Show Less
EINECSNumber 226-709-7
TSCA TSCA

RUO – Research Use Only