Learn More
Methyl 5-bromopyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6670603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl 5-bromopyrazine-2-carboxylate | |
| C6H5BrN2O2 | |
| 1 g | |
| PVTAJWAGBTWOPG-UHFFFAOYSA-N | |
| methyl 5-bromopyrazine-2-carboxylate | |
| 27282430 | |
| 97% |
| 210037-58-4 | |
| MFCD09991715 | |
| 5-bromo-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyrazinecarboxylic acid methyl ester, methyl5-bromopyrazine-2-carboxylate, ksc545m4b, 5-bromopyrazinecarboxylic acid methyl ester, pyrazinecarboxylicacid,5-bromo-,methylester, methyl 2-bromo-5-pyrazine-carboxylate, 2-pyrazinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrazinecarboxylicacid, 5-bromo-, methyl ester | |
| COC(=O)C1=CN=C(Br)C=N1 | |
| 217.02 | |
| 217.02 | |
| Solid |
Chemical Identifiers
| 210037-58-4 | |
| 217.02 | |
| PVTAJWAGBTWOPG-UHFFFAOYSA-N | |
| 27282430 | |
| COC(=O)C1=CN=C(Br)C=N1 |
| C6H5BrN2O2 | |
| MFCD09991715 | |
| 5-bromo-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyrazinecarboxylic acid methyl ester, methyl5-bromopyrazine-2-carboxylate, ksc545m4b, 5-bromopyrazinecarboxylic acid methyl ester, pyrazinecarboxylicacid,5-bromo-,methylester, methyl 2-bromo-5-pyrazine-carboxylate, 2-pyrazinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrazinecarboxylicacid, 5-bromo-, methyl ester | |
| methyl 5-bromopyrazine-2-carboxylate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only