missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromopyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7057241
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66706MD 250 mg
AAH6670603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66706MD Supplier Thermo Scientific Chemicals Supplier No. H66706MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 210037-58-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD09991715
InChI Key PVTAJWAGBTWOPG-UHFFFAOYSA-N
Synonym 5-bromo-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyrazinecarboxylic acid methyl ester, methyl5-bromopyrazine-2-carboxylate, ksc545m4b, 5-bromopyrazinecarboxylic acid methyl ester, pyrazinecarboxylicacid,5-bromo-,methylester, methyl 2-bromo-5-pyrazine-carboxylate, 2-pyrazinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrazinecarboxylicacid, 5-bromo-, methyl ester
PubChem CID 27282430
IUPAC Name methyl 5-bromopyrazine-2-carboxylate
SMILES COC(=O)C1=CN=C(Br)C=N1

Specifications

Quantity 250 mg
Formula Weight 217.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 5-bromopyrazine-2-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only