Learn More
Methyl 5-bromo-2-methylbenzoate, 98%
CAS: 79669-50-4 | C9H9BrO2 | 229.073 g/mol
Supplier: Thermo Scientific Chemicals H6186114
Description
Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| Methyl 5-bromo-2-methylbenzoate | |
| Yellow | |
| MFCD09839555 | |
| methyl 5-bromo-2-methyl-benzoate, methyl 5-bromo-o-toluate, methyl5-bromo-2-methylbenzoate, 5-bromo-2-methyl-benzoic acid methyl ester, benzoic acid, 5-bromo-2-methyl-, methyl ester, 5-bromo-2-methylbenzoic acid methyl ester, 5-bromo-2-methylbenzoicacid, methyl 4-bromo-toluate, pubchem10936, acmc-1bhme | |
| FDCYLMYCHALQJR-UHFFFAOYSA-N | |
| methyl 5-bromo-2-methylbenzoate | |
| 316201 | |
| 98% |
| 79669-50-4 | |
| C9H9BrO2 | |
| 25 g | |
| Soluble in water. | |
| CC1=C(C=C(C=C1)Br)C(=O)OC | |
| 229.073 | |
| 229.07 | |
| Powder |
Chemical Identifiers
| 79669-50-4 | |
| 229.073 | |
| FDCYLMYCHALQJR-UHFFFAOYSA-N | |
| 316201 | |
| CC1=C(C=C(C=C1)Br)C(=O)OC |
| C9H9BrO2 | |
| MFCD09839555 | |
| methyl 5-bromo-2-methyl-benzoate, methyl 5-bromo-o-toluate, methyl5-bromo-2-methylbenzoate, 5-bromo-2-methyl-benzoic acid methyl ester, benzoic acid, 5-bromo-2-methyl-, methyl ester, 5-bromo-2-methylbenzoic acid methyl ester, 5-bromo-2-methylbenzoicacid, methyl 4-bromo-toluate, pubchem10936, acmc-1bhme | |
| methyl 5-bromo-2-methylbenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only