missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-methylbenzoate, 98%

Catalog No. p-7054016
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6186106 5 g
AAH6186114 25 g
AAH6186122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6186106 Supplier Thermo Scientific Chemicals Supplier No. H6186106

CAS: 79669-50-4 | C9H9BrO2 | 229.073 g/mol

Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 79669-50-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD09839555
InChI Key FDCYLMYCHALQJR-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-methyl-benzoate, methyl 5-bromo-o-toluate, methyl5-bromo-2-methylbenzoate, 5-bromo-2-methyl-benzoic acid methyl ester, benzoic acid, 5-bromo-2-methyl-, methyl ester, 5-bromo-2-methylbenzoic acid methyl ester, 5-bromo-2-methylbenzoicacid, methyl 4-bromo-toluate, pubchem10936, acmc-1bhme
PubChem CID 316201
IUPAC Name methyl 5-bromo-2-methylbenzoate
SMILES CC1=C(C=C(C=C1)Br)C(=O)OC

Specifications

Color Yellow
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 229.07
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 5-bromo-2-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only