missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Bromo-2-methyl-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. M28451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28451G 1 g
M28455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28451G Supplier TCI America Supplier No. M28451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 220514-28-3
Molecular Formula C9H8BrNO4
Molecular Weight (g/mol) 274.07
MDL Number MFCD07781280
InChI Key UPBDNPCJIJAMPI-UHFFFAOYSA-N
Synonym 5-Bromo-2-methyl-3-nitrobenzoic Acid Methyl Ester
PubChem CID 24727998
IUPAC Name methyl 5-bromo-2-methyl-3-nitrobenzoate
SMILES COC(=O)C1=CC(Br)=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 1 g
Formula Weight 274.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromo-2-methyl-3-nitrobenzoate