CAS RN 220514-28-3
IUPAC Name:
methyl 5-bromo-2-methyl-3-nitrobenzoate
Synonyms:
methyl 5-bromo-2-methyl-3-nitrobenzoate
Molecular Weight (g/mol):
274.07
Molecular Formula:
C9H8BrNO4
InChi Key:
UPBDNPCJIJAMPI-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Br)=CC(=C1C)[N+]([O-])=O
1
–
1
of
1
results
1
Methyl 5-Bromo-2-methyl-3-nitrobenzoate 98.0+%, TCI America™
CAS: 220514-28-3 Molecular Formula: C9H8BrNO4 Molecular Weight (g/mol): 274.07 MDL Number: MFCD07781280 InChI Key: UPBDNPCJIJAMPI-UHFFFAOYSA-N Synonym: 5-Bromo-2-methyl-3-nitrobenzoic Acid Methyl Ester PubChem CID: 24727998 IUPAC Name: methyl 5-bromo-2-methyl-3-nitrobenzoate SMILES: COC(=O)C1=CC(Br)=CC(=C1C)[N+]([O-])=O