missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Amino-2-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7133280
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25405G 5 g
M254025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25405G Supplier TCI America Supplier No. M25405G

Chemical Identifiers

CAS 18595-12-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD08752568
InChI Key JNPZKGOLYSCSEL-UHFFFAOYSA-N
Synonym 5-amino-2-methyl-benzoic acid methyl ester, 5-amino-2-methylbenzoic acid methyl ester, 5-amino-2-methyl benzoic acid methyl ester, benzoic acid,5-amino-2-methyl-, methyl ester, benzoic acid, 5-amino-2-methyl-, methyl ester, 2-methyl-5-aminobenzoicacidmethylester, methyl 5-amino-o-toluate, methyl 2-methyl-5-aminobenzoate, methyl 3-amino-6-methylbenzoate, methyl 5-amino-2-methyl-benzoate
PubChem CID 15049977
IUPAC Name methyl 5-amino-2-methylbenzoate
SMILES COC(=O)C1=C(C)C=CC(N)=C1

Specifications

Melting Point 42°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Amino-2-methylbenzoate
TSCA No
Recommended Storage Refrigerator