missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Trifluoromethyl)benzoate 98.0+%, TCI America™

Catalog No. M14985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14985G 5 g
M149825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14985G Supplier TCI America Supplier No. M14985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2967-66-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042324
InChI Key VAZWXPJOOFSNLB-UHFFFAOYSA-N
Synonym methyl 4-trifluoromethyl benzoate, methyl-4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid methyl ester, methyl-4-trifluoromethylbenzoate, 3-cf3-c6h4-cooch3, methyl p-trifluoromethyl benzoate, methyl alpha,alpha,alpha-trifluoro-p-toluate, benzoic acid, 4-trifluoromethyl-, methyl ester, pubchem14120, acmc-209hab
PubChem CID 520447
IUPAC Name methyl 4-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-(Trifluoromethyl)benzoate