accessibility menu, dialog, popup

UserName

CH3OOFFF

CAS RN 2967-66-0

IUPAC Name: methyl 4-(trifluoromethyl)benzoate
Synonyms: methyl 4-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: VAZWXPJOOFSNLB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)