missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(trifluoromethyl)benzoate, 98%

CAS: 2967-66-0 | C9H7F3O2 | 204.15 g/mol

Supplier:  Thermo Scientific Chemicals A1540014

 View more versions of this product

Catalog No. AAA1540014


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 4-(trifluoromethyl)benzoate
13°C to 14°C
86°C to 87°C (10 mmHg)
C9H7F3O2
MFCD00042324
1963288
VAZWXPJOOFSNLB-UHFFFAOYSA-N
methyl 4-(trifluoromethyl)benzoate
520447
98%
2967-66-0
1.268
82°C (179°F)
1.452
25 g
methyl 4-trifluoromethyl benzoate, methyl-4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid methyl ester, methyl-4-trifluoromethylbenzoate, 3-cf3-c6h4-cooch3, methyl p-trifluoromethyl benzoate, methyl alpha,alpha,alpha-trifluoro-p-toluate, benzoic acid, 4-trifluoromethyl-, methyl ester, pubchem14120, acmc-209hab
COC(=O)C1=CC=C(C=C1)C(F)(F)F
204.15
204.15

Chemical Identifiers

2967-66-0
204.15
VAZWXPJOOFSNLB-UHFFFAOYSA-N
520447
COC(=O)C1=CC=C(C=C1)C(F)(F)F
C9H7F3O2
MFCD00042324
methyl 4-trifluoromethyl benzoate, methyl-4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid methyl ester, methyl-4-trifluoromethylbenzoate, 3-cf3-c6h4-cooch3, methyl p-trifluoromethyl benzoate, methyl alpha,alpha,alpha-trifluoro-p-toluate, benzoic acid, 4-trifluoromethyl-, methyl ester, pubchem14120, acmc-209hab
methyl 4-(trifluoromethyl)benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only