missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(trifluoromethyl)benzoate, 98%

Catalog No. AAA1540006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1540006 5 g
AAA1540014 25 g
AAA1540022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1540006 Supplier Thermo Scientific Chemicals Supplier No. A1540006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2967-66-0 | C9H7F3O2 | 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2967-66-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042324
InChI Key VAZWXPJOOFSNLB-UHFFFAOYSA-N
Synonym methyl 4-trifluoromethyl benzoate, methyl-4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid methyl ester, methyl-4-trifluoromethylbenzoate, 3-cf3-c6h4-cooch3, methyl p-trifluoromethyl benzoate, methyl alpha,alpha,alpha-trifluoro-p-toluate, benzoic acid, 4-trifluoromethyl-, methyl ester, pubchem14120, acmc-209hab
PubChem CID 520447
IUPAC Name methyl 4-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 13°C to 14°C
Density 1.268
Boiling Point 86°C to 87°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.452
Quantity 5 g
Beilstein 1963288
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only