missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-tert-Butylbenzoate 98.0+%, TCI America™

Catalog No. M169325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M169325G 25 g
M1693500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M169325G Supplier TCI America Supplier No. M169325G

Chemical Identifiers

CAS 26537-19-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
InChI Key UPIJOAFHOIWPLT-UHFFFAOYSA-N
Synonym methyl 4-tert-butyl benzoate, methyl p-tert-butylbenzoate, 4-tert-butylbenzoic acid methyl ester, unii-8vk4o27jmt, benzoic acid, 4-1,1-dimethylethyl-, methyl ester, methyl 4-t-butylbenzoate, 8vk4o27jmt, methyl4-tert-butylbenzoate, methyl p-t-butylbenzoate, benzoic acid, p-tert-butyl-, methyl ester
PubChem CID 97433
IUPAC Name methyl 4-tert-butylbenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OC

Specifications

Color Colorless
Boiling Point 247°C
Quantity 25 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-tert-Butylbenzoate
EINECSNumber (3)-4345
TSCA Yes