accessibility menu, dialog, popup

UserName

CH3OOCH3H3CCH3

CAS RN 26537-19-9

IUPAC Name: methyl 4-tert-butylbenzoate
Synonyms: methyl 4-T-butylbenzoate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: UPIJOAFHOIWPLT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(C)(C)C
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)