missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-tert-butylbenzoate, 98+%

Catalog No. AAA1861914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1861914 25 g
AAA1861922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1861914 Supplier Thermo Scientific Chemicals Supplier No. A1861914

CAS: 26537-19-9 | C12H16O2 | 192.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26537-19-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00008835
InChI Key UPIJOAFHOIWPLT-UHFFFAOYSA-N
Synonym methyl 4-tert-butyl benzoate, methyl p-tert-butylbenzoate, 4-tert-butylbenzoic acid methyl ester, unii-8vk4o27jmt, benzoic acid, 4-1,1-dimethylethyl-, methyl ester, methyl 4-t-butylbenzoate, 8vk4o27jmt, methyl4-tert-butylbenzoate, methyl p-t-butylbenzoate, benzoic acid, p-tert-butyl-, methyl ester
PubChem CID 97433
IUPAC Name methyl 4-tert-butylbenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OC

Specifications

Density 0.995
Boiling Point 122°C to 124°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.51
Quantity 25 g
Formula Weight 192.26
Percent Purity ≥98%
Chemical Name or Material Methyl 4-tert-butylbenzoate
EINECSNumber 247-768-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only