missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methylsalicylate 98.0+%, TCI America™

Catalog No. m15845g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M15845G 5 g
M158425G 25 g
2 options

Catalog No. M15845G

Supplier: TCI America M15845G

Chemical Identifiers

CAS 4670-56-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00020130
InChI Key UITFCFWKYAOJEJ-UHFFFAOYSA-N
Synonym methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate
PubChem CID 78400
IUPAC Name methyl 2-hydroxy-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)O

Specifications

Color White
Boiling Point 242°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Methyl 4-Methylsalicylate