accessibility menu, dialog, popup

UserName

CH3OOCH3HO

CAS RN 4670-56-8

IUPAC Name: methyl 2-hydroxy-4-methylbenzoate
Synonyms: methyl methylsalicylate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: UITFCFWKYAOJEJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C=C1O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)