missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methylsalicylate, 98+%

Catalog No. p-7056051
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6479306 5 g
AAH6479314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6479306 Supplier Thermo Scientific Chemicals Supplier No. H6479306

CAS: 4670-56-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4670-56-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00020130
InChI Key UITFCFWKYAOJEJ-UHFFFAOYSA-N
Synonym methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate
PubChem CID 78400
IUPAC Name methyl 2-hydroxy-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)O

Specifications

Melting Point 27°C to 28°C
Color Brown to Yellow
Density 1.147 g/mL
Boiling Point 242°C (literature)
Refractive Index 1.537
Quantity 5 g
Beilstein 2690247
Formula Weight 166.18
Percent Purity ≥98%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 4-methylsalicylate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-117-6
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only