missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methyl-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. M21465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21465G 5 g
M214625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21465G Supplier TCI America Supplier No. M21465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7356-11-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00130054
InChI Key YFPBHPCMYFCRKS-UHFFFAOYSA-N
Synonym methyl 3-nitro-4-methylbenzoate, 4-methyl-3-nitrobenzoic acid methyl ester, benzoic acid, 4-methyl-3-nitro-, methyl ester, methyl 3-nitro-p-toluate, 4-methyl-3-nitro-benzoic acid methyl ester, pubchem10934, acmc-1bfci, ksc497g4t, methyl4-methyl-3-nitrobenzoate, 4-methoxycarbonyl-2-nitrotoluene
PubChem CID 81816
IUPAC Name methyl 4-methyl-3-nitrobenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-]

Specifications

Melting Point 51°C
Color Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Methyl-3-nitrobenzoate